The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001770.5 | NP_001761.3 | 12 - 13 | 1552 | 68 |
XM_017023893.1 | XP_016879382.1 | 11 - 12 | 1353 | 68 | |
GenBank mRNA | AK130657.1 | - | 11 - 12 | 938 | 68 |
AK301167.1 | - | 12 - 13 | 1470 | 68 | |
AK304456.1 | - | 13 - 14 | 1337 | 68 | |
BC006338.2 | - | 12 - 13 | 1534 | 68 | |
M21097.1 | - | 12 - 13 | 1505 | 68 | |
X13312.1 | - | 12 - 13 | 1504 | 68 |