Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD19
Entrez Gene ID
930
Gene Name
CD19 molecule
Gene Aliases
B4, CVID3
Chromosome Location
Chr.16: 28931735 - 28939347 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178098.1 NP_001171569.1 2 - 3 416 67
NM_001770.5 NP_001761.3 2 - 3 416 67
XM_011545981.1 XP_011544283.1 2 - 3 416 67
GenBank mRNA AK301167.1 - 2 - 3 334 67
AK304456.1 - 3 - 4 201 67
AK313577.1 - 2 - 3 387 67
BC006338.2 - 2 - 3 398 67
M21097.1 - 2 - 3 370 67
X13312.1 - 2 - 3 369 67

View Full Product Details