The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317799.1 | NP_001304728.1 | 5 - 6 | 1213 | 113 |
NM_145207.2 | NP_660208.2 | 5 - 6 | 1216 | 113 | |
XM_005262783.4 | XP_005262840.1 | 5 - 6 | 1213 | 113 | |
XM_011531678.2 | XP_011529980.1 | 5 - 6 | 1213 | 113 | |
XM_011531679.2 | XP_011529981.1 | 5 - 6 | 1171 | 113 | |
XM_017007825.1 | XP_016863314.1 | 5 - 6 | 1212 | 113 | |
XM_017007826.1 | XP_016863315.1 | 5 - 6 | 1212 | 113 | |
XM_017007827.1 | XP_016863316.1 | 5 - 6 | 1212 | 113 | |
XM_017007828.1 | XP_016863317.1 | 5 - 6 | 1178 | 113 | |
XM_017007829.1 | XP_016863318.1 | 3 - 4 | 1836 | 113 | |
XM_017007830.1 | XP_016863319.1 | 5 - 6 | 1212 | 113 | |
GenBank mRNA | AF361489.1 | - | 5 - 6 | 1212 | 113 |
AF479656.1 | - | 5 - 6 | 1186 | 113 | |
AK091384.1 | - | 5 - 6 | 1171 | 113 | |
AL832175.1 | - | 5 - 6 | 1209 | 113 | |
BC048217.1 | - | 5 - 6 | 1156 | 113 |