The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024551.2 | NP_078827.2 | 5 - 6 | 840 | 138 |
XM_005253789.1 | XP_005253846.1 | 5 - 6 | 962 | 138 | |
XM_006719018.2 | XP_006719081.1 | 6 - 7 | 959 | 138 | |
XM_011521024.2 | XP_011519326.1 | 6 - 7 | 970 | 138 | |
GenBank mRNA | AK025085.1 | - | 2 - 3 | 368 | 138 |
AK127196.1 | - | 6 - 7 | 895 | 138 | |
AK128511.1 | - | 5 - 6 | 894 | 138 | |
AY424280.1 | - | 4 - 5 | 719 | 138 | |
BC051858.2 | - | 5 - 6 | 840 | 138 |