The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003150.3 | NP_003141.2 | 1 - 2 | 221 | 155 |
XM_005257617.3 | XP_005257674.2 | 1 - 2 | 229 | 155 | |
XM_017024972.1 | XP_016880461.1 | 1 - 2 | 229 | 155 | |
XM_017024974.1 | XP_016880463.1 | 1 - 2 | 229 | 155 | |
GenBank mRNA | AK301200.1 | - | 1 - 2 | 231 | 155 |
BC000627.2 | - | 1 - 2 | 140 | 155 | |
L29277.1 | - | 1 - 2 | 223 | 155 |