Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAT3
Entrez Gene ID
6774
Gene Name
signal transducer and activator of transcription 3
Gene Aliases
ADMIO, ADMIO1, APRF, HIES
Chromosome Location
Chr.17: 42313324 - 42388505 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003150.3 NP_003141.2 2 - 3 348 66
NM_139276.2 NP_644805.1 2 - 3 370 66
NM_213662.1 NP_998827.1 2 - 3 262 66
XM_005257616.3 XP_005257673.2 2 - 3 361 66
XM_005257617.3 XP_005257674.2 2 - 3 356 66
XM_011525145.2 XP_011523447.1 2 - 3 361 66
XM_011525146.2 XP_011523448.1 2 - 3 361 66
XM_017024972.1 XP_016880461.1 2 - 3 356 66
XM_017024973.1 XP_016880462.1 2 - 3 361 66
XM_017024974.1 XP_016880463.1 2 - 3 356 66
XM_017024975.1 XP_016880464.1 2 - 3 349 66
XM_017024976.1 XP_016880465.1 2 - 3 349 66
GenBank mRNA AB451232.1 - 1 - 2 130 66
AJ012463.1 - 1 - 2 130 66
AK291933.1 - 2 - 3 443 66
AK301200.1 - 2 - 3 358 66
AK316139.1 - 2 - 3 380 66
BC000627.2 - 2 - 3 267 66
BC014482.1 - 2 - 3 262 66
BI461226.1 - 2 - 3 375 66
L29277.1 - 2 - 3 350 66

View Full Product Details