Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AFF3
Entrez Gene ID
3899
Gene Name
AF4/FMR2 family member 3
Gene Aliases
LAF4, MLLT2-like
Chromosome Location
Chr.2: 99545861 - 100142583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025108.1 NP_001020279.1 17 - 18 3137 90
NM_002285.2 NP_002276.2 17 - 18 3154 90
XM_005263943.3 XP_005264000.2 16 - 17 3031 90
XM_005263945.3 XP_005264002.1 8 - 9 2388 90
XM_011511169.2 XP_011509471.1 17 - 18 3134 90
XM_011511170.2 XP_011509472.1 18 - 19 3273 90
XM_011511171.2 XP_011509473.1 17 - 18 3137 90
XM_011511172.2 XP_011509474.1 16 - 17 3089 90
XM_011511173.2 XP_011509475.1 17 - 18 3150 90
XM_011511174.2 XP_011509476.1 17 - 18 3094 90
XM_011511175.2 XP_011509477.1 19 - 20 3384 90
XM_011511176.2 XP_011509478.1 16 - 17 3075 90
XM_011511177.2 XP_011509479.1 17 - 18 3165 90
XM_011511178.2 XP_011509480.1 7 - 8 1896 90
XM_011511179.2 XP_011509481.1 7 - 8 1893 90
XM_017004085.1 XP_016859574.1 8 - 9 2385 90
XM_017004086.1 XP_016859575.1 8 - 9 2866 90
XM_017004087.1 XP_016859576.1 8 - 9 2863 90
GenBank mRNA AB209634.1 - 4 - 5 856 90
BC136579.1 - 15 - 16 2937 90
BC144266.1 - 16 - 17 3012 90
U34360.1 - 15 - 16 2972 90

View Full Product Details