The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020381.3 | NP_065114.3 | 3 - 4 | 921 | 157 |
XM_011535956.2 | XP_011534258.1 | 3 - 4 | 1019 | 157 | |
XM_011535957.2 | XP_011534259.1 | 3 - 4 | 1029 | 157 | |
XM_011535958.2 | XP_011534260.1 | 2 - 3 | 885 | 157 | |
XM_011535959.2 | XP_011534261.1 | 3 - 4 | 994 | 157 | |
XM_011535960.2 | XP_011534262.1 | 3 - 4 | 648 | 157 | |
XM_011535962.2 | XP_011534264.1 | 3 - 4 | 563 | 157 | |
XM_017011082.1 | XP_016866571.1 | 3 - 4 | 941 | 157 | |
GenBank mRNA | AB210839.1 | - | 3 - 4 | 631 | 157 |
AF254956.1 | - | 3 - 4 | 892 | 157 | |
AK225629.1 | - | 3 - 4 | 902 | 157 | |
AK301480.1 | - | 3 - 4 | 524 | 157 | |
AK315804.1 | - | 3 - 4 | 902 | 157 | |
BC039906.1 | - | 3 - 4 | 907 | 157 |