The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278784.1 | NP_001265713.1 | 4 - 5 | 470 | 85 |
NM_005827.2 | NP_005818.2 | 4 - 5 | 495 | 85 | |
XM_005256929.3 | XP_005256986.1 | 5 - 6 | 410 | 85 | |
XM_006721632.3 | XP_006721695.1 | 5 - 6 | 614 | 85 | |
XM_011524179.2 | XP_011522481.1 | 3 - 4 | 380 | 85 | |
XM_011524180.2 | XP_011522482.1 | 4 - 5 | 346 | 85 | |
GenBank mRNA | AK293563.1 | - | 4 - 5 | 495 | 85 |
AK296073.1 | - | 4 - 5 | 464 | 85 | |
AU309730.1 | - | 4 - 5 | 505 | 86 | |
BC011888.2 | - | 4 - 5 | 474 | 85 | |
CR745911.1 | - | 2 - 3 | 79 | 85 | |
CX164572.1 | - | 2 - 3 | 102 | 85 | |
D87989.1 | - | 4 - 5 | 464 | 85 |