Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HERPUD2
Entrez Gene ID
64224
Gene Name
HERPUD family member 2
Gene Aliases
-
Chromosome Location
Chr.7: 35632659 - 35695187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022373.4 NP_071768.3 8 - 9 1722 67
XM_006715765.1 XP_006715828.1 6 - 7 1530 67
XM_011515473.1 XP_011513775.1 5 - 6 1728 67
XM_017012506.1 XP_016867995.1 7 - 8 1920 67
GenBank mRNA AK025966.1 - 3 - 4 578 67
AK055708.1 - 7 - 8 1869 67
AK315685.1 - 7 - 8 1346 67
BC005091.1 - 8 - 9 1528 67
BC020264.1 - 8 - 9 1679 67

View Full Product Details