The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017434.4 | NP_059130.2 | 5 - 6 | 651 | 63 |
NM_175940.2 | NP_787954.1 | 4 - 5 | 459 | 63 | |
XM_011521681.2 | XP_011519983.1 | 3 - 4 | 352 | 63 | |
GenBank mRNA | AF213465.1 | - | 4 - 5 | 459 | 63 |
AF230495.1 | - | 5 - 6 | 651 | 63 | |
AK075268.1 | - | 4 - 5 | 459 | 63 | |
AK290372.1 | - | 5 - 6 | 712 | 63 | |
BC114628.1 | - | 4 - 5 | 448 | 63 | |
BC114938.1 | - | 4 - 5 | 448 | 63 |