Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG12
Entrez Gene ID
9140
Gene Name
autophagy related 12
Gene Aliases
APG12, APG12L, FBR93, HAPG12
Chromosome Location
Chr.5: 115828196 - 115841851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277783.1 NP_001264712.1 2 - 3 528 91
NM_004707.3 NP_004698.3 3 - 4 665 91
NR_033362.1 4 - 5 782 91
NR_033363.1 4 - 5 781 91
NR_073603.1 5 - 6 898 91
GenBank mRNA AB002440.1 - 2 - 3 126 91
AB017507.1 - 3 - 4 385 91
AK098251.1 - 3 - 4 1835 91
AK313320.1 - 3 - 4 660 91
AL161968.1 - 1 - 2 1753 91
BC011033.1 - 2 - 3 246 91
BC012266.2 - 3 - 4 616 91
CB995198.1 - 4 - 5 493 91
CN400030.1 - 4 - 5 416 91
CR978239.1 - 5 - 6 591 91
DB048378.1 - 2 - 3 236 91
EU554057.1 - 3 - 4 507 91

View Full Product Details