Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAGE1
Entrez Gene ID
8712
Gene Name
PAGE family member 1
Gene Aliases
AL5, CT16.3, GAGE-9, GAGEB1, PAGE-1
Chromosome Location
Chr.X: 49687450 - 49696019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003785.3 NP_003776.2 1 - 2 128 132
GenBank mRNA AF058989.1 - 1 - 2 124 131
BC004861.2 - 1 - 2 101 132
BU941413.1 - 1 - 2 130 132

View Full Product Details