The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002947.4 | NP_002938.1 | 7 - 8 | 1452 | 136 |
GenBank mRNA | AI243725.1 | - | 1 - 2 | 136 | |
AK311842.1 | - | 3 - 4 | 333 | 136 | |
BC005264.1 | - | 3 - 4 | 325 | 136 | |
BC009868.2 | - | 7 - 8 | 1452 | 136 | |
BT007320.1 | - | 3 - 4 | 280 | 136 | |
L07493.1 | - | 3 - 4 | 310 | 136 |