Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB2
Entrez Gene ID
3300
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Gene Aliases
CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3
Chromosome Location
Chr.2: 219279318 - 219286900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039550.1 NP_001034639.1 2 - 3 303 112
NM_006736.5 NP_006727.2 2 - 3 303 112
GenBank mRNA AK289626.1 - 2 - 3 126 112
AK292761.1 - 2 - 3 131 112
AK293888.1 - 2 - 3 255 112
AK312723.1 - 2 - 3 310 112
AL547521.3 - 2 - 3 349 112
BC011609.2 - 2 - 3 245 112
BC047056.1 - 1 - 2 106 112
BT007088.1 - 1 - 2 66 112
BX420085.2 - - 305 112
S37374.1 - 1 - 2 91 112
S37375.1 - 1 - 2 91 112

View Full Product Details