Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GMFG
Entrez Gene ID
9535
Gene Name
glia maturation factor gamma
Gene Aliases
GMF-GAMMA
Chromosome Location
Chr.19: 39328357 - 39336086 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301008.1 NP_001287937.1 2 - 3 160 82
NM_004877.3 NP_004868.1 3 - 4 257 82
GenBank mRNA AB001993.1 - 2 - 3 157 82
AF038956.1 - 2 - 3 151 82
AV714415.1 - 2 - 3 91 82
BC022207.1 - 3 - 4 177 82
BC032819.2 - 2 - 3 144 82
BC080180.1 - 3 - 4 177 82
BC093799.1 - 3 - 4 210 82
BC101818.1 - 3 - 4 210 82
BC143543.1 - 3 - 4 210 82
BC143546.1 - 3 - 4 209 82
BC143548.1 - 3 - 4 208 82
BF974742.1 - 3 - 4 258 82
CR456967.1 - 2 - 3 147 82
CR542114.1 - 2 - 3 147 82
EB387540.1 - 3 - 4 356 82

View Full Product Details