Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNX2
Entrez Gene ID
860
Gene Name
runt related transcription factor 2
Gene Aliases
AML3, CBF-alpha-1, CBFA1, CCD, CCD1, CLCD, OSF-2, OSF2, PEA2aA, PEBP2aA
Chromosome Location
Chr.6: 45328142 - 45664032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015051.3 NP_001015051.3 6 - 7 1061 117
NM_001024630.3 NP_001019801.3 6 - 7 1061 117
NM_001278478.1 NP_001265407.1 4 - 5 809 117
XM_011514960.2 XP_011513262.1 6 - 7 1236 117
XM_011514961.2 XP_011513263.1 6 - 7 1236 117
XM_011514962.2 XP_011513264.1 6 - 7 1236 117
XM_011514964.2 XP_011513266.1 6 - 7 1236 117
XM_011514965.2 XP_011513267.1 6 - 7 1236 117
XM_011514966.2 XP_011513268.1 4 - 5 1879 117
XM_017011391.1 XP_016866880.1 6 - 7 1271 117
XM_017011392.1 XP_016866881.1 4 - 5 812 117
XM_017011393.1 XP_016866882.1 6 - 7 1236 117
GenBank mRNA BC108919.1 - 4 - 5 791 117
BC108920.1 - 4 - 5 795 117
L40992.1 - 4 - 5 674 117

View Full Product Details