The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135673.2 | NP_001129145.1 | 11 - 12 | 1234 | 82 |
NM_001308076.1 | NP_001295005.1 | 11 - 12 | 1501 | 82 | |
NM_022374.3 | NP_071769.2 | 11 - 12 | 1234 | 82 | |
NR_024191.1 | 10 - 11 | 989 | 82 | ||
XM_011533017.1 | XP_011531319.1 | 11 - 12 | 1234 | 82 | |
XM_011533018.2 | XP_011531320.1 | 11 - 12 | 1644 | 82 | |
XM_011533019.2 | XP_011531321.1 | 11 - 12 | 1652 | 82 | |
XM_011533020.1 | XP_011531322.1 | 11 - 12 | 1332 | 82 | |
XM_011533022.2 | XP_011531324.1 | 10 - 11 | 984 | 82 | |
XM_011533023.2 | XP_011531325.1 | 11 - 12 | 2519 | 82 | |
XM_017004686.1 | XP_016860175.1 | 11 - 12 | 1197 | 82 | |
XM_017004687.1 | XP_016860176.1 | 11 - 12 | 2379 | 82 | |
XM_017004688.1 | XP_016860177.1 | 11 - 12 | 2519 | 82 | |
GenBank mRNA | AK026946.1 | - | 11 - 12 | 1217 | 82 |
AK225190.1 | - | 11 - 12 | 1220 | 82 | |
AK294049.1 | - | 11 - 12 | 1195 | 82 | |
AK294711.1 | - | 10 - 11 | 950 | 82 | |
AK302621.1 | - | 11 - 12 | 1501 | 82 | |
AK308894.1 | - | 10 - 11 | 974 | 82 | |
BC053508.1 | - | 10 - 11 | 961 | 82 | |
BX507348.1 | - | 4 - 5 | 293 | 82 |