The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017551.2 | NP_060021.1 | 12 - 13 | 2097 | 91 |
XM_011539720.2 | XP_011538022.1 | 12 - 13 | 2094 | 91 | |
GenBank mRNA | AB033046.1 | - | 9 - 10 | 1344 | 91 |
AK127168.1 | - | 12 - 13 | 2082 | 91 | |
AK293104.1 | - | 12 - 13 | 2055 | 91 | |
AK302192.1 | - | 5 - 6 | 869 | 91 | |
BC039263.1 | - | 12 - 13 | 2097 | 91 |