Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERLEC1
Entrez Gene ID
27248
Gene Name
endoplasmic reticulum lectin 1
Gene Aliases
C2orf30, CIM, CL24936, CL25084, HEL117, XTP3-B, XTP3TPB
Chromosome Location
Chr.2: 53786931 - 53826928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127398.2 NP_001120870.1 10 - 11 1344 126
NM_015701.4 NP_056516.2 11 - 12 1506 126
XM_011532766.1 XP_011531068.1 11 - 12 1420 126
GenBank mRNA AF131743.1 - 3 - 4 325 126
AF131849.1 - 5 - 6 695 126
AK315477.1 - 11 - 12 1295 126
AY358717.1 - 10 - 11 1228 126
AY453410.1 - 11 - 12 1357 126
BC013129.1 - 11 - 12 1357 126
BC022228.1 - 10 - 11 1192 126
EU794676.1 - 11 - 12 1506 126

View Full Product Details