Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERLEC1
Entrez Gene ID
27248
Gene Name
endoplasmic reticulum lectin 1
Gene Aliases
C2orf30, CIM, CL24936, CL25084, HEL117, XTP3-B, XTP3TPB
Chromosome Location
Chr.2: 53786931 - 53826928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127398.2 NP_001120870.1 11 - 12 1423 96
NM_015701.4 NP_056516.2 12 - 13 1585 96
XM_011532766.1 XP_011531068.1 12 - 13 1499 96
GenBank mRNA AF131743.1 - 4 - 5 404 96
AF131849.1 - 6 - 7 774 96
AK001913.1 - 12 - 13 1466 96
AK315477.1 - 12 - 13 1374 96
AY358717.1 - 11 - 12 1307 96
AY453410.1 - 12 - 13 1436 96
BC013129.1 - 12 - 13 1436 96
BC022228.1 - 11 - 12 1271 96
EU794676.1 - 12 - 13 1585 96

View Full Product Details