Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERLEC1
Entrez Gene ID
27248
Gene Name
endoplasmic reticulum lectin 1
Gene Aliases
C2orf30, CIM, CL24936, CL25084, HEL117, XTP3-B, XTP3TPB
Chromosome Location
Chr.2: 53786931 - 53826928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127397.2 NP_001120869.1 9 - 10 1320 152
NM_015701.4 NP_056516.2 9 - 10 1320 152
XM_011532766.1 XP_011531068.1 9 - 10 1234 152
GenBank mRNA AF131743.1 - 1 - 2 139 152
AF131849.1 - 3 - 4 509 152
AK001913.1 - 9 - 10 1201 152
AK315477.1 - 9 - 10 1109 152
AY453410.1 - 9 - 10 1171 152
BC013129.1 - 9 - 10 1171 152
BQ017438.1 - 4 - 5 152
EU794676.1 - 9 - 10 1320 152

View Full Product Details