The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322178.1 | NP_001309107.1 | 10 - 11 | 2054 | 101 |
NM_001322179.1 | NP_001309108.1 | 11 - 12 | 2204 | 101 | |
NM_001322180.1 | NP_001309109.1 | 10 - 11 | 2010 | 101 | |
NM_001322181.1 | NP_001309110.1 | 11 - 12 | 2201 | 101 | |
NM_001322187.1 | NP_001309116.1 | 12 - 13 | 2269 | 101 | |
NM_001322188.1 | NP_001309117.1 | 11 - 12 | 2204 | 101 | |
NM_001322189.1 | NP_001309118.1 | 12 - 13 | 2463 | 101 | |
NM_001322190.1 | NP_001309119.1 | 10 - 11 | 2060 | 101 | |
NM_001322191.1 | NP_001309120.1 | 10 - 11 | 2005 | 101 | |
NM_033401.4 | NP_207837.2 | 11 - 12 | 2204 | 101 | |
NM_138994.4 | NP_620481.2 | 10 - 11 | 1598 | 101 | |
NR_136210.1 | 11 - 12 | 2204 | 101 | ||
NR_136213.1 | 11 - 12 | 2199 | 101 | ||
NR_136216.1 | 13 - 14 | 2387 | 101 | ||
XM_011523403.2 | XP_011521705.1 | 11 - 12 | 1742 | 101 | |
XM_017023805.1 | XP_016879294.1 | 11 - 12 | 1933 | 101 | |
GenBank mRNA | AB051550.2 | - | 11 - 12 | 2143 | 101 |
AB100092.1 | - | 10 - 11 | 1614 | 101 | |
AB100093.1 | - | 9 - 10 | 1539 | 101 | |
AB100094.1 | - | 11 - 12 | 1758 | 101 | |
AF463518.1 | - | 10 - 11 | 1906 | 101 | |
AK057327.1 | - | 11 - 12 | 1728 | 101 |