Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ABI2
- Entrez Gene ID
-
10152
- Gene Name
- abl-interactor 2
- Gene Aliases
-
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
- Chromosome Location
-
Chr.2: 203327874 - 203432173 on Build GRCh38
View Full Product Details