The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017003119.1 | XP_016858608.1 | 10 - 11 | 1914 | 115 |
XM_017003122.1 | XP_016858611.1 | 9 - 10 | 1767 | 115 | |
XM_017003126.1 | XP_016858615.1 | 8 - 9 | 1713 | 115 | |
GenBank mRNA | AK299092.1 | - | 7 - 8 | 686 | 115 |