Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPH9
Entrez Gene ID
221421
Gene Name
radial spoke head 9 homolog
Gene Aliases
C6orf206, CILD12, MRPS18AL1
Chromosome Location
Chr.6: 43645030 - 43671011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193341.1 NP_001180270.1 5 - 6 793 73
XM_005248901.3 XP_005248958.1 5 - 6 811 73
GenBank mRNA AK055407.1 - 5 - 6 733 73

View Full Product Details