Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELMOD3
Entrez Gene ID
84173
Gene Name
ELMO domain containing 3
Gene Aliases
DFNB88, LST3, RBED1, RBM29
Chromosome Location
Chr.2: 85354394 - 85391752 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK307252.1 - 2 - 3 228 86

View Full Product Details