The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032380.4 | NP_115756.2 | 2 - 3 | 508 | 147 |
NM_170681.2 | NP_733781.1 | 2 - 3 | 508 | 147 | |
NM_170691.2 | NP_733792.1 | 2 - 3 | 508 | 147 | |
NR_104006.1 | 2 - 3 | 508 | 147 | ||
XM_011543691.2 | XP_011541993.1 | 2 - 3 | 291 | 147 | |
GenBank mRNA | AF111808.1 | - | 2 - 3 | 258 | 147 |
AF367997.1 | - | 2 - 3 | 354 | 147 | |
AK025314.1 | - | 2 - 3 | 285 | 147 | |
AK307839.1 | - | 2 - 3 | 198 | 147 | |
AK308422.1 | - | 2 - 3 | 255 | 147 | |
AK315705.1 | - | 2 - 3 | 287 | 147 | |
AY453587.1 | - | 2 - 3 | 252 | 147 | |
BC030612.1 | - | 2 - 3 | 257 | 147 | |
BI828937.1 | - | 2 - 3 | 511 | 147 | |
DC402212.1 | - | 2 - 3 | 505 | 147 |