The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145415.1 | NP_001138887.1 | 13 - 14 | 2401 | 76 |
NM_012432.3 | NP_036564.3 | 13 - 14 | 2401 | 76 | |
XM_005245641.3 | XP_005245698.1 | 13 - 14 | 2404 | 76 | |
XM_017002953.1 | XP_016858442.1 | 13 - 14 | 2328 | 76 | |
GenBank mRNA | BC028671.2 | - | 13 - 14 | 2328 | 76 |
D31891.1 | - | 13 - 14 | 2297 | 76 |