The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145415.1 | NP_001138887.1 | 7 - 8 | 1066 | 72 |
NM_001243491.1 | NP_001230420.1 | 7 - 8 | 1012 | 72 | |
NM_012432.3 | NP_036564.3 | 7 - 8 | 1066 | 72 | |
XM_005245641.3 | XP_005245698.1 | 7 - 8 | 1066 | 72 | |
XM_017002953.1 | XP_016858442.1 | 7 - 8 | 990 | 72 | |
XM_017002954.1 | XP_016858443.1 | 7 - 8 | 1066 | 72 | |
XM_017002955.1 | XP_016858444.1 | 7 - 8 | 1074 | 72 | |
GenBank mRNA | AK302461.1 | - | 3 - 4 | 571 | 72 |
AK308335.1 | - | 7 - 8 | 1010 | 72 | |
BC009362.2 | - | 6 - 7 | 902 | 72 | |
BC028671.2 | - | 7 - 8 | 993 | 72 | |
D31891.1 | - | 7 - 8 | 962 | 72 |