The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171162.1 | NP_001164633.1 | 17 - 18 | 3566 | 69 |
NM_005096.3 | NP_005087.1 | 17 - 18 | 3041 | 69 | |
NM_201599.2 | NP_963893.1 | 17 - 18 | 3032 | 69 | |
XM_005262309.3 | XP_005262366.1 | 17 - 18 | 3475 | 69 | |
XM_005262310.2 | XP_005262367.1 | 17 - 18 | 2952 | 69 | |
XM_011531062.2 | XP_011529364.1 | 17 - 18 | 2941 | 69 | |
GenBank mRNA | AB002383.1 | - | 17 - 18 | 2940 | 69 |
BC069057.1 | - | 17 - 18 | 2945 | 69 | |
X95808.1 | - | 18 - 19 | 3542 | 69 |