Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGNBP2
Entrez Gene ID
79893
Gene Name
gametogenetin binding protein 2
Gene Aliases
DIF-3, DIF3, LCRG1, LZK1, ZFP403, ZNF403
Chromosome Location
Chr.17: 36544888 - 36589848 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024835.4 NP_079111.1 12 - 13 1959 123
XM_005257689.2 XP_005257746.1 12 - 13 1941 123
XM_011525265.2 XP_011523567.1 11 - 12 1704 123
XM_011525266.1 XP_011523568.1 11 - 12 1827 123
XM_017025107.1 XP_016880596.1 11 - 12 1672 123
GenBank mRNA AF126964.1 - 11 - 12 1690 123
AJ404670.2 - 4 - 5 516 123
AK026214.1 - 4 - 5 593 123
AK290687.1 - 11 - 12 1715 123
AK301614.1 - 9 - 10 1247 123
BC137491.1 - 12 - 13 1825 123

View Full Product Details