The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024835.4 | NP_079111.1 | 4 - 5 | 747 | 135 |
XM_005257689.2 | XP_005257746.1 | 4 - 5 | 723 | 135 | |
XM_011525265.2 | XP_011523567.1 | 3 - 4 | 486 | 135 | |
XM_011525266.1 | XP_011523568.1 | 4 - 5 | 723 | 135 | |
XM_017025107.1 | XP_016880596.1 | 3 - 4 | 460 | 135 | |
GenBank mRNA | AF126964.1 | - | 3 - 4 | 478 | 135 |
AF268387.1 | - | 4 - 5 | 667 | 135 | |
AK024883.1 | - | 4 - 5 | 703 | 135 | |
AK290687.1 | - | 3 - 4 | 503 | 135 | |
AK292161.1 | - | 4 - 5 | 716 | 135 | |
BC137491.1 | - | 4 - 5 | 613 | 135 |