The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024835.4 | NP_079111.1 | 6 - 7 | 964 | 126 |
XM_005257689.2 | XP_005257746.1 | 6 - 7 | 940 | 126 | |
XM_011525265.2 | XP_011523567.1 | 5 - 6 | 703 | 126 | |
XM_017025107.1 | XP_016880596.1 | 5 - 6 | 677 | 126 | |
GenBank mRNA | AF126964.1 | - | 5 - 6 | 695 | 126 |
AF268387.1 | - | 6 - 7 | 884 | 126 | |
AK024883.1 | - | 6 - 7 | 920 | 126 | |
AK290687.1 | - | 5 - 6 | 720 | 126 | |
AK292161.1 | - | 6 - 7 | 933 | 126 | |
BC137491.1 | - | 6 - 7 | 830 | 126 |