The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318758.1 | NP_001305687.1 | 7 - 8 | 1745 | 81 |
NM_002504.5 | NP_002495.2 | 7 - 8 | 1745 | 81 | |
NM_147134.3 | NP_667345.1 | 7 - 8 | 1745 | 81 | |
GenBank mRNA | AF332009.1 | - | 7 - 8 | 1650 | 81 |
AK225750.1 | - | 7 - 8 | 1618 | 81 | |
AK291643.1 | - | 7 - 8 | 1635 | 81 | |
AL832342.1 | - | 7 - 8 | 1628 | 81 | |
BC006447.1 | - | 6 - 7 | 1242 | 81 | |
BC012151.1 | - | 7 - 8 | 1615 | 81 | |
U15306.1 | - | 6 - 7 | 1541 | 81 |