The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318758.1 | NP_001305687.1 | 9 - 10 | 2056 | 83 |
NM_002504.5 | NP_002495.2 | 9 - 10 | 2056 | 83 | |
NM_147134.3 | NP_667345.1 | 9 - 10 | 2056 | 83 | |
GenBank mRNA | AF332009.1 | - | 9 - 10 | 1961 | 83 |
AK225750.1 | - | 9 - 10 | 1929 | 83 | |
AK291643.1 | - | 9 - 10 | 1946 | 83 | |
AL832342.1 | - | 9 - 10 | 1939 | 83 | |
BC006447.1 | - | 8 - 9 | 1553 | 83 | |
BC012151.1 | - | 9 - 10 | 1926 | 83 | |
U15306.1 | - | 8 - 9 | 1852 | 83 |