The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002766.2 | NP_002757.2 | 2 - 3 | 661 | 92 |
GenBank mRNA | AK131332.1 | - | 2 - 3 | 662 | 92 |
AK298171.1 | - | 2 - 3 | 127 | 92 | |
AK312637.1 | - | 2 - 3 | 372 | 92 | |
BC009012.1 | - | 2 - 3 | 140 | 92 |