Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRPSAP1
Entrez Gene ID
5635
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 1
Gene Aliases
PAP39
Chromosome Location
Chr.17: 76310787 - 76354149 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002766.2 NP_002757.2 5 - 6 1027 82
XM_011525034.2 XP_011523336.1 4 - 5 725 82
GenBank mRNA AK131332.1 - 5 - 6 1028 82
AK298171.1 - 5 - 6 493 82
AK312637.1 - 5 - 6 738 82
BC009012.1 - 5 - 6 506 82
D61391.1 - 6 - 7 543 82

View Full Product Details