The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040006.1 | NP_001035095.1 | 10 - 11 | 1793 | 146 |
NM_006706.3 | NP_006697.2 | 11 - 12 | 1856 | 146 | |
XM_005268365.2 | XP_005268422.1 | 11 - 12 | 2042 | 197 | |
XM_017008977.1 | XP_016864466.1 | 10 - 11 | 1979 | 197 | |
XM_017008978.1 | XP_016864467.1 | 11 - 12 | 2042 | 197 | |
XM_017008979.1 | XP_016864468.1 | 11 - 12 | 1855 | 146 | |
XM_017008980.1 | XP_016864469.1 | 11 - 12 | 2042 | 197 | |
XM_017008981.1 | XP_016864470.1 | 9 - 10 | 1448 | 146 | |
XM_017008982.1 | XP_016864471.1 | 10 - 11 | 1773 | 197 | |
XM_017008983.1 | XP_016864472.1 | 10 - 11 | 1773 | 146 | |
GenBank mRNA | AB209910.1 | - | 10 - 11 | 1767 | 146 |
AF017789.1 | - | 11 - 12 | 1816 | 146 | |
AK126455.1 | - | 9 - 10 | 1696 | 146 | |
BC014151.2 | - | 10 - 11 | 1787 | 146 | |
BC111727.1 | - | 10 - 11 | 1771 | 146 |