The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040006.1 | NP_001035095.1 | 6 - 7 | 1376 | 147 |
NM_006706.3 | NP_006697.2 | 7 - 8 | 1439 | 147 | |
XM_005268365.2 | XP_005268422.1 | 7 - 8 | 1625 | 147 | |
XM_017008977.1 | XP_016864466.1 | 6 - 7 | 1562 | 147 | |
XM_017008978.1 | XP_016864467.1 | 7 - 8 | 1625 | 147 | |
XM_017008979.1 | XP_016864468.1 | 7 - 8 | 1438 | 147 | |
XM_017008980.1 | XP_016864469.1 | 7 - 8 | 1625 | 147 | |
XM_017008981.1 | XP_016864470.1 | 5 - 6 | 1031 | 147 | |
XM_017008982.1 | XP_016864471.1 | 6 - 7 | 1356 | 147 | |
XM_017008983.1 | XP_016864472.1 | 6 - 7 | 1356 | 147 | |
XM_017008984.1 | XP_016864473.1 | 7 - 8 | 1625 | 147 | |
XM_017008985.1 | XP_016864474.1 | 7 - 8 | 1625 | 147 | |
GenBank mRNA | AB209910.1 | - | 6 - 7 | 1350 | 147 |
AF017789.1 | - | 7 - 8 | 1399 | 147 | |
AK304319.1 | - | 6 - 7 | 1375 | 147 | |
BC014151.2 | - | 6 - 7 | 1370 | 147 | |
BC111727.1 | - | 6 - 7 | 1354 | 147 |