The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001397.2 | NP_001388.1 | 1 - 2 | 131 | 62 |
GenBank mRNA | AK290656.1 | - | 1 - 2 | 103 | 62 |
AK296486.1 | - | 1 - 2 | 106 | 62 | |
AK309677.1 | - | 1 - 2 | 105 | 62 | |
BC117256.1 | - | 1 - 2 | 131 | 62 | |
BC126257.1 | - | 1 - 2 | 131 | 62 | |
D49471.1 | - | 1 - 2 | 56 | 62 | |
Z35307.1 | - | 1 - 2 | 42 | 62 |