The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003198.2 | NP_003189.2 | 3 - 4 | 376 | 70 |
GenBank mRNA | AF116729.1 | - | 3 - 4 | 225 | 70 |
AK310229.1 | - | 3 - 4 | 362 | 70 | |
AK313079.1 | - | 3 - 4 | 248 | 70 | |
BC002883.2 | - | 3 - 4 | 260 | 70 | |
DB461774.1 | - | 3 - 4 | 414 | 70 | |
L47345.1 | - | 3 - 4 | 270 | 70 |