The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003198.2 | NP_003189.2 | 4 - 5 | 1570 | 69 |
GenBank mRNA | AF116729.1 | - | 4 - 5 | 1419 | 69 |
AK310229.1 | - | 4 - 5 | 1556 | 69 | |
AK313079.1 | - | 4 - 5 | 1442 | 69 | |
BC002883.2 | - | 4 - 5 | 1454 | 69 | |
BC020448.1 | - | 1 - 2 | 182 | 69 | |
L47345.1 | - | 4 - 5 | 1464 | 69 |