The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007220.3 | NP_009151.1 | 1 - 2 | 68 | 71 |
GenBank mRNA | AB021660.1 | - | 1 - 2 | 88 | 71 |
AK291050.1 | - | 1 - 2 | 87 | 71 | |
BP371510.1 | - | 1 - 2 | 68 | 71 |