Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAK
Entrez Gene ID
2580
Gene Name
cyclin G associated kinase
Gene Aliases
DNAJ26, DNAJC26
Chromosome Location
Chr.4: 849275 - 932390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318134.1 NP_001305063.1 8 - 9 1174 125
NM_005255.3 NP_005246.2 11 - 12 1411 125
XM_005272268.2 XP_005272325.1 11 - 12 1351 125
XM_005272270.2 XP_005272327.1 11 - 12 1351 125
XM_011513425.2 XP_011511727.1 11 - 12 1351 125
XM_011513426.2 XP_011511728.1 11 - 12 1351 125
XM_011513427.2 XP_011511729.1 10 - 11 1261 125
XM_011513428.2 XP_011511730.1 11 - 12 1351 125
XM_011513429.2 XP_011511731.1 9 - 10 1673 125
XM_011513430.1 XP_011511732.1 8 - 9 1098 125
XM_011513431.2 XP_011511733.1 11 - 12 1351 125
XM_011513432.2 XP_011511734.1 11 - 12 1322 125
XM_011513434.2 XP_011511736.1 9 - 10 1163 125
XM_017007991.1 XP_016863480.1 10 - 11 1261 125
XM_017007992.1 XP_016863481.1 7 - 8 1008 125
XM_017007993.1 XP_016863482.1 9 - 10 1100 125
XM_017007994.1 XP_016863483.1 11 - 12 1351 125
XM_017007995.1 XP_016863484.1 10 - 11 1170 125
GenBank mRNA AB208854.1 - 8 - 9 125
AK299554.1 - 8 - 9 1046 125
AK309413.1 - 8 - 9 1030 125
BC085005.1 - 8 - 9 1157 125
D88435.1 - 11 - 12 1208 125

View Full Product Details