Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GAK
- Entrez Gene ID
-
2580
- Gene Name
- cyclin G associated kinase
- Gene Aliases
-
DNAJ26, DNAJC26
- Chromosome Location
-
Chr.4: 849275 - 932390 on Build GRCh38
View Full Product Details