The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000109.3 | NP_000100.2 | 57 - 58 | 8869 | 124 |
NM_004006.2 | NP_003997.1 | 57 - 58 | 8793 | 124 | |
NM_004009.3 | NP_004000.1 | 57 - 58 | 8800 | 124 | |
NM_004010.3 | NP_004001.1 | 57 - 58 | 8883 | 124 | |
NM_004011.3 | NP_004002.2 | 29 - 30 | 4571 | 124 | |
NM_004012.3 | NP_004003.1 | 29 - 30 | 4714 | 124 | |
NM_004013.2 | NP_004004.1 | 14 - 15 | 2210 | 124 | |
NM_004014.2 | NP_004005.1 | 3 - 4 | 423 | 124 | |
NM_004020.3 | NP_004011.2 | 14 - 15 | 2210 | 124 | |
NM_004021.2 | NP_004012.1 | 14 - 15 | 2210 | 124 | |
NM_004022.2 | NP_004013.1 | 14 - 15 | 2210 | 124 | |
NM_004023.2 | NP_004014.1 | 14 - 15 | 2210 | 124 | |
XM_006724468.2 | XP_006724531.1 | 57 - 58 | 8746 | 124 | |
XM_006724469.3 | XP_006724532.1 | 57 - 58 | 8720 | 124 | |
XM_006724470.3 | XP_006724533.1 | 57 - 58 | 8746 | 124 | |
XM_006724473.2 | XP_006724536.1 | 56 - 57 | 8608 | 124 | |
XM_006724474.3 | XP_006724537.1 | 57 - 58 | 8746 | 124 | |
XM_006724475.2 | XP_006724538.1 | 57 - 58 | 8746 | 124 | |
XM_011545467.1 | XP_011543769.1 | 56 - 57 | 8623 | 124 | |
XM_011545468.2 | XP_011543770.1 | 57 - 58 | 8746 | 124 | |
XM_017029328.1 | XP_016884817.1 | 57 - 58 | 8746 | 124 | |
XM_017029331.1 | XP_016884820.1 | 17 - 18 | 3322 | 124 | |
GenBank mRNA | AB208836.1 | - | 11 - 12 | 1657 | 124 |
BC127103.1 | - | 11 - 12 | 1734 | 124 | |
BC150141.1 | - | 11 - 12 | 1734 | 124 | |
M18533.1 | - | 57 - 58 | 8757 | 124 | |
X14298.1 | - | 57 - 58 | 8647 | 124 |