The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000109.3 | NP_000100.2 | 59 - 60 | 9253 | 85 |
NM_004006.2 | NP_003997.1 | 59 - 60 | 9177 | 85 | |
NM_004009.3 | NP_004000.1 | 59 - 60 | 9184 | 85 | |
NM_004010.3 | NP_004001.1 | 59 - 60 | 9267 | 85 | |
NM_004011.3 | NP_004002.2 | 31 - 32 | 4955 | 85 | |
NM_004012.3 | NP_004003.1 | 31 - 32 | 5098 | 85 | |
NM_004013.2 | NP_004004.1 | 16 - 17 | 2594 | 85 | |
NM_004014.2 | NP_004005.1 | 5 - 6 | 807 | 85 | |
NM_004020.3 | NP_004011.2 | 16 - 17 | 2594 | 85 | |
NM_004021.2 | NP_004012.1 | 16 - 17 | 2594 | 85 | |
NM_004022.2 | NP_004013.1 | 16 - 17 | 2594 | 85 | |
NM_004023.2 | NP_004014.1 | 16 - 17 | 2594 | 85 | |
XM_006724468.2 | XP_006724531.1 | 59 - 60 | 9130 | 85 | |
XM_006724469.3 | XP_006724532.1 | 59 - 60 | 9104 | 85 | |
XM_006724470.3 | XP_006724533.1 | 59 - 60 | 9130 | 85 | |
XM_006724473.2 | XP_006724536.1 | 58 - 59 | 8992 | 85 | |
XM_006724474.3 | XP_006724537.1 | 59 - 60 | 9130 | 85 | |
XM_006724475.2 | XP_006724538.1 | 59 - 60 | 9130 | 85 | |
XM_011545467.1 | XP_011543769.1 | 58 - 59 | 9007 | 85 | |
XM_011545468.2 | XP_011543770.1 | 59 - 60 | 9130 | 85 | |
XM_017029328.1 | XP_016884817.1 | 59 - 60 | 9130 | 85 | |
XM_017029331.1 | XP_016884820.1 | 19 - 20 | 3706 | 85 | |
GenBank mRNA | AB208836.1 | - | 13 - 14 | 2041 | 85 |
BC127103.1 | - | 13 - 14 | 2118 | 85 | |
BC150141.1 | - | 13 - 14 | 2118 | 85 | |
M18533.1 | - | 59 - 60 | 9141 | 85 | |
X14298.1 | - | 59 - 60 | 9031 | 85 |