The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004646.3 | NP_004637.1 | 17 - 18 | 2491 | 56 |
GenBank mRNA | AF035835.1 | - | 17 - 18 | 2335 | 56 |
AF190637.1 | - | 17 - 18 | 2335 | 56 | |
EU642886.1 | - | 17 - 18 | 2335 | 56 | |
EU642887.1 | - | 17 - 18 | 2334 | 56 |