The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004646.3 | NP_004637.1 | 18 - 19 | 2668 | 121 |
GenBank mRNA | AF035835.1 | - | 18 - 19 | 2512 | 121 |
AF190637.1 | - | 18 - 19 | 2512 | 121 | |
EU642886.1 | - | 18 - 19 | 2512 | 121 | |
EU642887.1 | - | 18 - 19 | 2511 | 121 |